Learn More
Perkin Elmer US LLC Green Injection Port Septa, Pkg. 10
Catalog No. 502104645
Encompass_Preferred
The result is a mid-range general purpose septum made of uniquely formulated silicone rubber you can use for all your daily analyses. These septa also benefit from also with the non-stick coating and the CenterGuide design.
Catalog No. 50-210-4645 Supplier Perkin Elmer US LLC Supplier No. N9306218
The result is a mid-range general purpose septum made of uniquely formulated silicone rubber you can use for all your daily analyses. These septa also benefit from also with the non-stick coating and the CenterGuide design.