Learn More
Perkin Elmer US LLC Green Injection Port Septa, Pkg. 50
Catalog No. 502104644
Encompass_Preferred
The result is a mid-range general purpose septum made of uniquely formulated silicone rubber you can use for all your daily analyses. These septa also benefit from also with the non-stick coating and the CenterGuide design.
Catalog No. 50-210-4644 Supplier Perkin Elmer US LLC Supplier No. N9306219
The result is a mid-range general purpose septum made of uniquely formulated silicone rubber you can use for all your daily analyses. These septa also benefit from also with the non-stick coating and the CenterGuide design.