Learn More
Cole-Parmer Keck Standard Taper Clips, Joint Size 24, Green, Pk/12
Catalog No. NC9843933 Shop All Cole-Parmer Products
Encompass_Preferred
These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).
This item is not returnable. View return policy
Catalog No. NC9843933 Supplier Cole-Parmer Supplier No. 0684015
These economical standard taper clips securely hold conical joints under pressure without damaging glass. The polyoxymethylene (POM) acetal resin clips are safe to 302°>F (150°>C). Resistant to concentrated alkalies and dilute acids (not resistant to halogen gases and concentrated acids).