Learn More
Edmund Optics GREEN LASER POINTER
Catalog No. NC9159706
Encompass
Ultra-Bright Green Laser Pointer, Visible up to 6000 ft., Maximum Output Power: <5mW, Wavelength: 532nm, Beam Diameter: 1.1mm, Beam Divergence: <1.2mrad, Power Requirements: 2 AAA Batteries (included), Operating Time: 3 hr. continuous use
Catalog No. NC9159706 Supplier Edmund Optics Supplier No. 55789
Ultra-Bright Green Laser Pointer, Visible up to 6000 ft., Maximum Output Power: <5mW, Wavelength: 532nm, Beam Diameter: 1.1mm, Beam Divergence: <1.2mrad, Power Requirements: 2 AAA Batteries (included), Operating Time: 3 hr. continuous use