Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Green light-emitting spiro-copolymer

Catalog No. 900441250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-044-1250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-044-1250MG Supplier Sigma Aldrich Supplier No. 900441250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Features:
  • Soluble green light-emitting spiro-copolymer.
  • High efficiency.
  • Long lifetime, with further potential to improve.
  • Saturated green color coordinates, with low operation voltage.
  • Typical performance measured in standard device stack (ITO/PEDOT:PSS 20 nm/SPG-01T 80 nm/Ba 6 nm/Ag 150 nm):
  • Max. efficiency: 11 Cd/A.
  • V_on: 2.5 V.
  • V at 100 cd/m2: 3.6 V.
  • V at 1000 cd/m2: 6.0 V.
  • Max. power eff.: 12 lm/W.
  • External quantum eff.: 4.1%.
  • CIE1931 at 100 cd/m2: (0.33, 0.59).
  • DC lifetime at 100 cd/m2 at RT: >10k hrs.
  • AC lifetime at 200 cd/m2 at RT: >5000 hrs.Product of EMD Performance Materials Corp., an Affiliate of Merck KGaA, Darmstadt, Germany

Specifications

CAS 1430803-21-6
Quantity 250 mg
Synonym Livilux(TM) SPG-01 T
Molecular Weight (g/mol) 100000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.