Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Earth Force™ Low-Cost Estuary Monitoring Kit
SDPSCI_ED

Catalog No. S65403 Shop All LaMotte Company Products
Change view
Click to view available options
pH Range:
4 to 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
S65403 4 to 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S65403 Supplier LaMotte™ Supplier No. 591120
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Manual (in English and Spanish) with diagrammed instructions, laminated color chart, all necessary apparatus and TesTabs reagents

Simple, nonhazardous methods to test saline and brackish waters found in estuaries.

  • Test water samples for coliform bacteria, salinity, dissolved oxygen, nitrate, pH, phosphate, temperature and turbidity
  • Supplied with all TesTabs™ reagents and components to test 10 water samples (two samples for coliform)

Specifications

pH Range 4 to 10
Volume (Metric) 40 oz.
For Use With (Application) To test saline and brackish waters found in estuaries
Includes Manual with diagrammed instructions, laminated color chart, all necessary apparatus and testabs™ reagents
Type Green Low Cost Estuary Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.