Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Earth Force™ Low-Cost Water Monitoring Kit
SDPSCI_ED

Catalog No. S65401 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
4+
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S65401 4+
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S65401 Supplier LaMotte™ Supplier No. 588620
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Manual (in English and Spanish) with diagrammed instructions, laminated color chart, all necessary apparatus and TesTabs reagents

Learn the basics of water quality monitoring and testing.

  • Analyze sample water for pH, dissolved oxygen, biochemical oxygen demand, temperature, turbidity, nitrate, phosphate and coliform bacteria
  • Each nonhazardous, TesTabs™ reagent tests 10 water samples (three samples for coliform)

Order Info

Grades 4+.

Specifications

pH Range 4 to 10
For Use With (Application) Water quality monitoring and testing
Grade 4+
Includes Manual (in English and Spanish) with diagrammed instructions, laminated color chart, all necessary apparatus and testabs™ reagents
Type Environmental Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.