Learn More
Montana Molecular Mitochondria-Targeted mNeonGreen (Green MitoPaint) Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Catalog No. NC3787516
Encompass_Preferred
Bright, green fluorescent mitochondrial label for live-cell imaging—no fixation, staining, or cell lysis required. Visualize mitochondrial morphology and dynamics in real time, over hours to days, in living cells. Delivered via Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Targeted to mitochondria via Cox8. Ideal for dynamic, non-destructive studies of mitochondrial organization, cellular phenotypes, and high-content imaging.
Catalog No. NC3787516 Supplier Montana Molecular Supplier No. P1010G005
Bright, green fluorescent mitochondrial label for live-cell imaging—no fixation, staining, or cell lysis required. Visualize mitochondrial morphology and dynamics in real time, over hours to days, in living cells. Delivered via Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Targeted to mitochondria via Cox8. Ideal for dynamic, non-destructive studies of mitochondrial organization, cellular phenotypes, and high-content imaging.