Learn More
HORIZON DISCOVERY BIOSCIENCES LIMITED Accell Green Non-targeting Control siRNA, 20nmol
Catalog No. NC1617805
Encompass
Accell Green Non-targeting Control siRNA provides qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA or FACS analysis to determine general uptake. Non-targeting siRNA reagents are designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses.
Catalog No. NC1617805 Supplier HORIZON DISCOVERY BIOSCIENCES LIMITED Supplier No. D0019500120
Accell Green Non-targeting Control siRNA provides qualitative assessment of Accell siRNA uptake as determined by fluorescent microscopy to evaluate cytoplamic localization of the dye-labeled Accell siRNA or FACS analysis to determine general uptake. Non-targeting siRNA reagents are designed to aid in the interpretation of experiments with Accell siRNA reagents as highly reliable controls for assessment of baseline cellular responses.