Learn More
Chemglass Life Sciences Cap, GL-45, Green, Open Top (1-5/16" Hole), Polypropylene
SDP
Catalog No. 501215139
Encompass
Open top polypropylene caps for use with port assemblies.Caps can be autoclaved in steam to 140 degreesC.
Catalog No. 50-121-5139 Supplier Chemglass Life Sciences Supplier No. CLS148122
Open top polypropylene caps for use with port assemblies.Caps can be autoclaved in steam to 140 degreesC.