Learn More
Next Advance Inc Green RINO RNA Lysis Kit, 250 pack, 1.5 mL tubes, RNase free, DNase free, stainless steel beads
SDP
Catalog No. NC3401808
Encompass
The Next Advance Green RINO RNA Lysis Kit provides an efficient solution for homogenizing small, tough samples to isolate high-quality RNA. This kit includes pre-filled RINO screw-cap tubes designed for seamless use with Bullet Blenders, ensuring robust sample preparation while maintaining RNA integrity for downstream applications. Each tube is pre-loaded with RNase and DNase-free stainless steel beads, reducing contamination risks during the sample preparation process.
  • Efficient homogenization of small, tough biological samples.
  • Certified RNase and DNase free for high-quality RNA isolation.
  • Pre-filled tubes save preparation time and reduce contamination risks.
  • Designed for compatibility with Bullet Blenders for reliable performance.
Catalog No. NC3401808 Supplier Next Advance Inc Supplier No. GREENR5RNA
The Next Advance Green RINO RNA Lysis Kit provides an efficient solution for homogenizing small, tough samples to isolate high-quality RNA. This kit includes pre-filled RINO screw-cap tubes designed for seamless use with Bullet Blenders, ensuring robust sample preparation while maintaining RNA integrity for downstream applications. Each tube is pre-loaded with RNase and DNase-free stainless steel beads, reducing contamination risks during the sample preparation process.
  • Efficient homogenization of small, tough biological samples.
  • Certified RNase and DNase free for high-quality RNA isolation.
  • Pre-filled tubes save preparation time and reduce contamination risks.
  • Designed for compatibility with Bullet Blenders for reliable performance.