Learn More
Nelson Jameson Company Green Hand Pad
Catalog No. NC0964808
Encompass
Light to medium duty and the perfect addition to any color-coded program. Open web design allows for quick washing and reuse. Pad conforms to cracks and crevices on the surface to get a deep down clean. Will not splinter or rust. Easy on hands. 6"W x 9"L. 20 pads/case. Not recommended for soft plastics, fine china, or stainless steel.
Catalog No. NC0964808 Supplier Nelson Jameson Company Supplier No. 4562960
Light to medium duty and the perfect addition to any color-coded program. Open web design allows for quick washing and reuse. Pad conforms to cracks and crevices on the surface to get a deep down clean. Will not splinter or rust. Easy on hands. 6"W x 9"L. 20 pads/case. Not recommended for soft plastics, fine china, or stainless steel.