Learn More
Bioworld GREEN SEPAROPOREAGAROSE)
Catalog No. 50197917
Encompass
GREEN SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consists of Reactive Green-19 bound to cross-linked 4% agarose beads (Separopore 4B-CL). Specifications Matrix: Cross-linked agarose, 4% Dye:Reactive Green 19 Extent o...
Catalog No. 50-197-917 Supplier Bioworld Supplier No. 201810992
GREEN SEPAROPORE (AGAROSE) 4B-CL: A dye-ligand affinity matrix consists of Reactive Green-19 bound to cross-linked 4% agarose beads (Separopore 4B-CL). Specifications Matrix: Cross-linked agarose, 4% Dye:Reactive Green 19 Extent o...