Learn More
Caresfield Green, Specimen Transport Bag
SDP
Catalog No. 21222124
Change view
Click to view available options
Color:
Green
Catalog No. Color
21-222-124 Green
1 options
Catalog No. 21-222-124 Supplier Caresfield Supplier No. 48609GREEN
Add to Cart
Add to Cart
6 x 9 in. Biohazard Specimen Bag 3-Wall Tear Away ZipTop Closure Green 1000/Cs.
  • Specimen transport size supports routine lab and clinical sample transfer workflows.
  • Biohazard specimen bag design with green color coding for quick visual identification and sorting.
  • Tear-away ZipTop closure enables secure sealing and controlled opening when processing samples.
  • Supports high-volume hospitals, clinics, diagnostic labs, and courier transport use.
Specifications
Autoclavable Not Autoclavable
Closure Type Ziplock
Color Green
Disposable Single-use
Material Plastic
Sterility Non-sterile
Latex Free Yes
Quantity 1000/Cs.
Length (English) 6 in.
Width (English) 9 in.
Product Type Zip Closure Bag