Learn More
INFORS USA INC Multitron, Incubator Shakers
SDP
Catalog No. NC1666704
Encompass
The Multitron is an incubator shaker by Infors HT. It has a temperature range of 5°C above ambient to 80°C, allowing incubation at physiological temperatures. Speed is adjustable from 25 to 400rpm with an orbit of 20mm. The inner chamber is made of corrosion resistant stainless steel and has a capacity of 2 to 150 liters. Safety features include overtemperature protection, imbalance detection, and door locking. It can accommodate flasks up to 5 liters and microplates. The Multitron allows gentle to vigorous shaking for microbial and cell cultures.
This item is not returnable. View return policy
Catalog No. NC1666704 Supplier INFORS USA INC Supplier No. 78113
The Multitron is an incubator shaker by Infors HT. It has a temperature range of 5°C above ambient to 80°C, allowing incubation at physiological temperatures. Speed is adjustable from 25 to 400rpm with an orbit of 20mm. The inner chamber is made of corrosion resistant stainless steel and has a capacity of 2 to 150 liters. Safety features include overtemperature protection, imbalance detection, and door locking. It can accommodate flasks up to 5 liters and microplates. The Multitron allows gentle to vigorous shaking for microbial and cell cultures.