Learn More
Genscript Corporation Green Taq DNA Polymerase
Catalog No. NC0868068
Encompass
Green Taq DNA Polymerase is designed to increase the stability of the Taq enzyme for more convenient transport and applications. The polymerase (1000 U) is designed for 400 rxns if 2.5 U are used per 50 ul PCR reaction. It can be stable for six months if stored at 4°C or for up to one month without significant loss of activity at ambient temperature. Our technology can increase the stability of Green Taq DNA at higher temperature (72°C). The higher yield PCR product can be harvested in long PCR amplification.
Catalog No. NC0868068 Supplier Genscript Corporation Supplier No. E00043
Green Taq DNA Polymerase is designed to increase the stability of the Taq enzyme for more convenient transport and applications. The polymerase (1000 U) is designed for 400 rxns if 2.5 U are used per 50 ul PCR reaction. It can be stable for six months if stored at 4°C or for up to one month without significant loss of activity at ambient temperature. Our technology can increase the stability of Green Taq DNA at higher temperature (72°C). The higher yield PCR product can be harvested in long PCR amplification.