Learn More
Montana Molecular Tau-Targeted mNeonGreen (Green TauPaint) Expression Kit - Big Sky BacMam, 5mL - Montana Molecular
SDP
Catalog No. NC3787549
Encompass_Preferred
Bright, green fluorescent protein fused to Tau for live-cell imaging—no fixation, staining, or cell lysis required. Visualize cytoskeletal structures and cell morphology, over hours to days, in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Ideal for non-destructive studies of Tau organization, cellular phenotypes, and high-content imaging.
Catalog No. NC3787549 Supplier Montana Molecular Supplier No. P1050G005
Bright, green fluorescent protein fused to Tau for live-cell imaging—no fixation, staining, or cell lysis required. Visualize cytoskeletal structures and cell morphology, over hours to days, in living cells. Delivered using Montana Molecular’s Big Sky BacMam vector for enhanced expression across a wide range of cell types, including primary cultures and iPSCs. Ideal for non-destructive studies of Tau organization, cellular phenotypes, and high-content imaging.