Learn More
Medchemexpress LLC Green Tea Extract | 25 G
SDP
Catalog No. 5000389616
Encompass_Preferred
Green Tea Extract is derived from Green Tea, containing 30% to 98% Tea Polyphenol. It is intended for research use only and not for sale to patients.
  • Available in various quantities: 5 g, 10 g, 25 g, 50 g, 100 g, and quantities greater than 100 g (available via quote).
  • Solid appearance, color ranges from brown to reddish brown.
  • Soluble in DMSO (100 mg/mL with sonication).
  • Can be stored at room temperature (dry and cool); in solvent, it can be stored at -80°C for 1 year or -20°C for 6 months.
Catalog No. 50-003-89616 Supplier Medchemexpress LLC Supplier No. 25GHYN13229
Green Tea Extract is derived from Green Tea, containing 30% to 98% Tea Polyphenol. It is intended for research use only and not for sale to patients.
  • Available in various quantities: 5 g, 10 g, 25 g, 50 g, 100 g, and quantities greater than 100 g (available via quote).
  • Solid appearance, color ranges from brown to reddish brown.
  • Soluble in DMSO (100 mg/mL with sonication).
  • Can be stored at room temperature (dry and cool); in solvent, it can be stored at -80°C for 1 year or -20°C for 6 months.