Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Green Tea Extract Mix, Reversed Phase, UV, MS, Solid

Catalog No. 502391240 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Green Tea Extract mix provides a reproducible background signal for evaluating LC and MS systems, particularly for small molecule applications where natural alternatives are impractical. This LC-MS standard is used in system benchmarking and adheres to strict quality control.

  • Provides reproducible background signal for LC and MS system evaluation.
  • Suitable for small molecule applications needing a complex matrix.
  • Verifies system performance before analyses.
  • Separation mode: Reversed phase.
  • Detection method: UV, MS.
  • Formulation: Solid.
  • Storage condition: Room temperature.
  • System type: LC, LCMS.
  • Shelf life: 60 months.
  • Mass: 33.33 mg.
  • Application: System performance, small molecule.
  • Units per package: 1 pk.

Catalog No. 50-239-1240 Supplier Waters Corp Supplier No. 186006962
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Green Tea Extract mix provides a reproducible background signal for evaluating LC and MS systems, particularly for small molecule applications where natural alternatives are impractical. This LC-MS standard is used in system benchmarking and adheres to strict quality control.

  • Provides reproducible background signal for LC and MS system evaluation.
  • Suitable for small molecule applications needing a complex matrix.
  • Verifies system performance before analyses.
  • Separation mode: Reversed phase.
  • Detection method: UV, MS.
  • Formulation: Solid.
  • Storage condition: Room temperature.
  • System type: LC, LCMS.
  • Shelf life: 60 months.
  • Mass: 33.33 mg.
  • Application: System performance, small molecule.
  • Units per package: 1 pk.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.