Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Qorpak™ Green Thermoset Cap with F217 and PTFE Liner

Catalog No. 02913106
Change view
Click to view available options
Neck Finish:
13-425
15-425
18-400
20-400
22-400
24-400
28-400
33-400
38-400
8-425
Product Type:
Lined Cap (Ribbed)
Lined Cap (Smooth)
Packaging:
Bagged
Bulk
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish Product Type Packaging
02-913-106 8-425 Lined Cap (Ribbed) Bagged
02-913-105 8-425 Lined Cap (Ribbed) Bulk
02-913-107 13-425 Lined Cap (Ribbed) Bagged
02-913-108 15-425 Lined Cap (Ribbed) Bulk
02-913-109 15-425 Lined Cap (Ribbed) Bagged
02-913-110 18-400 Lined Cap (Ribbed) Bulk
02-913-111 18-400 Lined Cap (Smooth) Bagged
02-913-112 20-400 Lined Cap (Smooth) Bulk
02-913-113 20-400 Lined Cap (Smooth) Bagged
02-913-114 22-400 Lined Cap (Smooth) Bulk
02-913-115 22-400 Lined Cap (Smooth) Bagged
02-913-116 24-400 Lined Cap (Smooth) Bulk
02-913-117 24-400 Lined Cap (Smooth) Bagged
02-913-118 28-400 Lined Cap (Smooth) Bulk
02-913-119 28-400 Lined Cap (Smooth) Bagged
02-913-120 33-400 Lined Cap (Smooth) Bulk
02-913-121 33-400 Lined Cap (Smooth) Bagged
02-913-123 38-400 Lined Cap (Smooth) Bagged
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 02-913-106 Supplier Qorpak™ Supplier No. 272635
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provides a wide range of chemical compatability and temperature tolerant. Qorpak™ Green Thermoset Cap with F217 and PTFE Liner are ideal for analytical lab samples, high purity chemicals, strong acids solvents, environmental samples, pharmaceuticals, and diagnostic reagents.

  • The color green easily identifies PTFE liner system
  • F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
  • Provides odor and impact resilience for a tight seal with a low moisture transmission rate
  • F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
  • Available in smooth and ribbed style

Specifications

Color Green
Material Thermoset
No. per Case 500
Packaging Bagged
Sterility Non-sterile
Thread Size (Metric) 72 mm
Liner Material F217, PTFE
Neck Finish 8-425
Autoclavable Not Autoclavable
Disposable Reusable
Product Type Lined Cap (Ribbed)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.