Learn More
Qorpak™ Green Thermoset Cap with F217 and PTFE Liner PROMO
Description
- The color green easily identifies PTFE liner system
- F-217 foam-backed PTFE liners resist attack from virtually all chemicals at room temperature
- Provides odor and impact resilience for a tight seal with a low moisture transmission rate
- F-217 has a low density foam core between two solid layers of LDPE (low density polyethylene)
- Available in smooth and ribbed style
Specifications
Specifications
| Color | Green |
| Material | Thermoset |
| No. per Case | 500/Cs. |
| Packaging | Bagged |
| Liner Material | F217, PTFE |
| Neck Finish | 24-400 |
| Closure Material | Thermoset |
| Product Type | Thermoset Cap |