Learn More
Andwin Scientific GREEN TOUGH-SPOTS LARGE 1/2
SDP
Catalog No. 50820986
Encompass_Preferred
The green Tough-Spots labels are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2".
Catalog No. 50-820-986 Supplier Andwin Scientific Supplier No. 91850502
The green Tough-Spots labels are specially designed to be used for 0.5 mL tubes. The labels are resistant against chemicals and temperatures from -196C to 80C. This product can be put on several surfaces like most plastics, glass, vinyl, and metals. Any marking instrument can also be utilized to write on these labels. This product has a diameter of 1/2".