Learn More
Eurofins Discoverx Green V-Bottom PP Ligand Dilution Plates, 10 plates/pk. (92-0011)
Catalog No. NC0921909
Encompass
These 96-well plates are green, V-bottomed, untreated and nonsterile plates. These plates are intended for use to dilute compounds to create dose response curves, and to mix reagents for assays. Contains 10 plates. For detailed product information, User manual and datasheet, visit www.discoverx.com/microplates.
Catalog No. NC0921909 Supplier Eurofins Discoverx Supplier No. 920011
These 96-well plates are green, V-bottomed, untreated and nonsterile plates. These plates are intended for use to dilute compounds to create dose response curves, and to mix reagents for assays. Contains 10 plates. For detailed product information, User manual and datasheet, visit www.discoverx.com/microplates.