Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Earth Force™ Standard Water Monitoring Kit
SDPSCI_ED

Catalog No. S65402 Shop All LaMotte Company Products
Change view
Click to view available options
Volume (Metric):
20 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
S65402 20 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65402 Supplier LaMotte™ Supplier No. 5848
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Set of nine test factor modules, each including: diagrammed instructions, color chart, test factor flashcard, all necessary apparatus and nonhazardous TesTabs™ reagents to perform 100 tests (44 tests for coliform bacteria)

An inquiry-based approach to watershed education.

  • Great tool for beginning a school-based, community-involved water monitoring program
  • Learn to design and implement a river study through the use of watershed maps
  • Compile and analyze results, share the results with your community and learn to take action through a variety of productive avenues

Order Info

Grades 4+

Specifications

pH Range 4 to 10
Volume (Metric) 20 oz.
For Use With (Application) Environmental education
Grade 4+
Includes Biochemical oxygen demand, coliform bacteria, dissolved oxygen, nitrate, pH, phosphate, turbidity, temperature, diagrammed instructions, color chart, test factor flashcard, all necessary apparatus and nonhazardous testabs™ reagents
Type Water Monitoring Kit
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.