Learn More
MSC Betco Degreaser, Green Earth Natural, 2 L Bottle, 4-Pack, Mandarin Zest
Catalog No. 502835842
Encompass
This degreaser is a naturally preferred solution for all degreasing tasks. It utilizes a synergistic blend of natural, biorenewable, and biobased ingredients with a unique surfactant system, providing cleaning power equal to traditional degreasers. It effectively breaks down greases, oils, and fats, including industrial petroleum greases, from a variety of water-safe surfaces.
  • Removes inks, oils, greases, and fats from various surfaces.
  • Offers cleaning performance comparable to traditional degreasers.
  • Formulated with a proprietary blend of natural, biorenewable, and biobased ingredients.
  • Breaks down greases, oils, and industrial petroleum greases on all water-safe surfaces.
  • Works with FastDraw dispensing options.
  • One 2 L bottle yields 17 gallons of solution.
  • Meets Green Earth criteria.
Catalog No. 50-283-5842 Supplier MSC Supplier No. 72526627
This degreaser is a naturally preferred solution for all degreasing tasks. It utilizes a synergistic blend of natural, biorenewable, and biobased ingredients with a unique surfactant system, providing cleaning power equal to traditional degreasers. It effectively breaks down greases, oils, and fats, including industrial petroleum greases, from a variety of water-safe surfaces.
  • Removes inks, oils, greases, and fats from various surfaces.
  • Offers cleaning performance comparable to traditional degreasers.
  • Formulated with a proprietary blend of natural, biorenewable, and biobased ingredients.
  • Breaks down greases, oils, and industrial petroleum greases on all water-safe surfaces.
  • Works with FastDraw dispensing options.
  • One 2 L bottle yields 17 gallons of solution.
  • Meets Green Earth criteria.