Learn More
MSC Sellars Wipers, Jumbo Roll, Natural, DRC, 950 Sheets
Catalog No. 502837746
Encompass
These eco-friendly and high-performance wipers offer a sustainable solution for everyday cleaning. Made with up to 60% recycled fibers, they are super-absorbent, tackle spills with ease, and remain strong even when wet. They are ideal for cleaning and disinfecting across various environments, including schools, universities, government facilities, and LEED-certified buildings.
  • Designed for light-duty wiping applications, such as removing oil and grease, or cleaning kitchen areas and countertops.
  • Jumbo rolls fit wall mount or floor stand dispensers, offering quick access for large-scale usage.
  • Remain strong when wet and are super-absorbent.
  • Meet EPA comprehensive procurement guidelines and can earn LEED-EB operation and maintenance credits due to recycled fiber content.
  • Manufactured in the U.S.A.
Catalog No. 50-283-7746 Supplier MSC Supplier No. 57522963
These eco-friendly and high-performance wipers offer a sustainable solution for everyday cleaning. Made with up to 60% recycled fibers, they are super-absorbent, tackle spills with ease, and remain strong even when wet. They are ideal for cleaning and disinfecting across various environments, including schools, universities, government facilities, and LEED-certified buildings.
  • Designed for light-duty wiping applications, such as removing oil and grease, or cleaning kitchen areas and countertops.
  • Jumbo rolls fit wall mount or floor stand dispensers, offering quick access for large-scale usage.
  • Remain strong when wet and are super-absorbent.
  • Meet EPA comprehensive procurement guidelines and can earn LEED-EB operation and maintenance credits due to recycled fiber content.
  • Manufactured in the U.S.A.