Learn More
Chemglass Life Sciences Gregar Extractor Body Only
SDP
Catalog No. CG137502
Encompass_Preferred
The Gregar Extractor† is the first major advance in laboratory extractor design in over 100 years. Developed by Joseph Gregar and Ken Anderson of Argonne National Laboratories, this extractor is the only unit on the market that can perform all three common types of extraction (See the illustrations). During solid extractions the extractor operates upon the principle of continuous solvent displacement, permitting the sample to be immersed in solvent at all times, thereby increasing the efficiency of the solid-solvent interactions. The design eliminates packing and/or channeling of solid samples normally associated with common soxhlet extractors. U.S. Patent Number D413678†Designed and developed by Joseph Gregar and Ken Anderson of Argonne National Laboratories
Catalog No. CG137502 Supplier Chemglass Life Sciences Supplier No. CG137502
The Gregar Extractor† is the first major advance in laboratory extractor design in over 100 years. Developed by Joseph Gregar and Ken Anderson of Argonne National Laboratories, this extractor is the only unit on the market that can perform all three common types of extraction (See the illustrations). During solid extractions the extractor operates upon the principle of continuous solvent displacement, permitting the sample to be immersed in solvent at all times, thereby increasing the efficiency of the solid-solvent interactions. The design eliminates packing and/or channeling of solid samples normally associated with common soxhlet extractors. U.S. Patent Number D413678†Designed and developed by Joseph Gregar and Ken Anderson of Argonne National Laboratories