Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One SNAPPY Short Standard Holder with Ridges

Catalog No. 22040376
This item is not returnable. View return policy
Catalog No. 22-040-376 Supplier Greiner Bio-One Supplier No. 450202

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Helps to resist pushback during blood collection

  • Two grips molded into the holder grasp the bottom of the VACUETTE blood collection tube safety cap to help resist pushback
  • Compatible with all blood collection tubes

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Product Type Short Standard Holder
Storage Requirements 4°C to 25°C
For Use With (Application) Helps to resist pushback during Blood collection
Certifications/Compliance CE, OSHA
Material Plastic
Includes Ridges
For Use With (Equipment) All Blood collection tubes
Latex Free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.