Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Trace Element Studies Evacuated Tube

Catalog No. 22040309
Change view
Click to view available options
Capacity (Metric):
6 mL
Closure Color:
Royal Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Closure Color
22040309 6 mL Royal Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22040309 Supplier Greiner Bio-One Supplier No. 456275
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Designed for testing of trace elements

  • For testing of trace elements such as aluminum, lead, cadmium, chrome, iron, fluoride, cobalt, copper, lithium, manganese, molybdenum, mercury, selenium or thalium
  • Contains sodium heparin to activate antithrombins, thus blocking the coagulation cascade and producing a whole blood/ plasma sample instead of clotted blood plus serum
  • Manufactured from virtually unbreakable clear PET plastic
  • Royal blue non-ridged pull caps with black exterior ring

TRUSTED_SUSTAINABILITY

Specifications

Additive NH Trace Elements Sodium Heparin
Sterility Sterile
Capacity (Metric) 6 mL
Closure Color Royal Blue
Length (Metric) 100 mm
Diameter (Metric) Outer 13 mm
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.