Learn More
Sigma Aldrich Fine Chemicals Biosciences Greiner UV-Star(R) 96 well plates flat bottom clear cyclic olefin copolymer (COC) wells (cycloolefine) | 40EA
Catalog No. NC2018170
Encompass
Greiner UV-Star(R) 96 well plates flat bottom clear cyclic olefin copolymer (COC) wells (cycloolefine) | 40EA
Catalog No. NC2018170 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M381240EA
Greiner UV-Star(R) 96 well plates flat bottom clear cyclic olefin copolymer (COC) wells (cycloolefine) | 40EA