Learn More
Greiner Bio-One HOLDEX™ plus Butterfly
Catalog No. p-183881
Change view
Click to view available options
:
Holdex + Butterfly: 19 cm tubing, 23G x 8/10in. needle
Holdex + Butterfly: 8 cm tubing, 23G x 8/10in. needle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
22-040-086 Holdex + Butterfly: 8 cm tubing, 23G x 8/10in. needle
22-040-087 Holdex + Butterfly: 19 cm tubing, 23G x 8/10in. needle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 22-040-086 Supplier Greiner Bio-One Supplier No. 450272
This item is not returnable. View return policy
A single-use holder with preattached, sterile butterflies
  • The offset Luer™ adapter of the Holdex tube ensures an optimal venipuncture angle which is advantageous for difficult vein conditions
  • Holdex and the attached butterfly are disposed of after use, reducing needlestick injuries