Learn More
Aviva Systems Biology Corp GREM2 Antibody - N-terminal region (ARP57635_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57635_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GREM2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GREM2. Alternate names: CKTSF1B2, DAND3, PRDCApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-62918