Learn More
ACROBiosystems ACROBiosystems Human Gremlin / GREM1 Protein, Fc Tag
Supplier:  ACROBiosystems GR1H52541MG
Encompass_Preferred
Recombinant Protein;1MG;Human Gremlin, Fc Tag (GR1-H5254) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 184 (Accession # NP_037504).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 44.2 kDa. The protein migrates as 48-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BMP-2, Tag Free (Cat. No. BM2-H4117) at 2μg/mL (100 μL/well) can bind Human Gremlin, Fc Tag (Cat. No. GR1-H5254) with a linear range of 0.8-80 ng/mL (QC tested).
Catalog No. 50-195-7411