Learn More
ACROBiosystems ACROBiosystems Human Gremlin / GREM1 Protein, His Tag
Catalog No. 501957417
Encompass_Preferred
Recombinant Protein;100UG;Human Gremlin, His Tag (GR1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 184 (Accession # NP_037504).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BMP-2, Tag Free (Cat. No. BM2-H4117) at 2 μg/mL (100 μL/well) can bind Human Gremlin, His Tag (Cat. No. GR1-H52H3) with a linear range of 0.02-0.313 μg/mL (QC tested).
Catalog No. 50-195-7417 Supplier ACROBiosystems Supplier No. GR1H52H3100UG
Recombinant Protein;100UG;Human Gremlin, His Tag (GR1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Lys 25 - Asp 184 (Accession # NP_037504).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 20.2 kDa. The protein migrates as 28-35 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human BMP-2, Tag Free (Cat. No. BM2-H4117) at 2 μg/mL (100 μL/well) can bind Human Gremlin, His Tag (Cat. No. GR1-H52H3) with a linear range of 0.02-0.313 μg/mL (QC tested).