Learn More
Medchemexpress LLC Grepafloxacin hydrochloride | 161967-81-3 | 99.55% | C19H23ClFN3O3 | 50 MG
SDP
Catalog No. 5000432639
Encompass_Preferred
Grepafloxacin hydrochloride, also known as OPC-17116 hydrochloride, is an orally active fluoroquinolone antibiotic. It demonstrates potent activity against common community-acquired respiratory pathogens, including *Streptococcus pneumoniae*. This compound is characterized by its high tissue penetration and a favorable pharmacodynamic profile, making it a promising agent for research into antibiotic treatments.
  • Potent activity against community-acquired respiratory pathogens.
  • Effective against *Streptococcus pneumoniae*.
  • Exhibits high tissue penetration.
  • Possesses a promising pharmacodynamic profile.
  • Demonstrates in vitro antibacterial activity against Gram-positive bacteria.
  • Shows in vivo efficacy in experimental systemic infections caused by Gram-positive and Gram-negative bacteria.
Catalog No. 50-004-32639 Supplier Medchemexpress LLC Supplier No. HYA0147A50MG
Grepafloxacin hydrochloride, also known as OPC-17116 hydrochloride, is an orally active fluoroquinolone antibiotic. It demonstrates potent activity against common community-acquired respiratory pathogens, including *Streptococcus pneumoniae*. This compound is characterized by its high tissue penetration and a favorable pharmacodynamic profile, making it a promising agent for research into antibiotic treatments.
  • Potent activity against community-acquired respiratory pathogens.
  • Effective against *Streptococcus pneumoniae*.
  • Exhibits high tissue penetration.
  • Possesses a promising pharmacodynamic profile.
  • Demonstrates in vitro antibacterial activity against Gram-positive bacteria.
  • Shows in vivo efficacy in experimental systemic infections caused by Gram-positive and Gram-negative bacteria.