Learn More
Bioworld Gresshoff and Doy (DBM2) Medium w/ Vitamins, 50 L
Catalog No. 50254617
Encompass_Preferred
Gresshoff & Doy (DBM2) MediumContains Macronutrients, Micronutrients, and Vitamins as described Gresshoff & Doy (1974)Working Concentration: 2.71 g/L Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested
Catalog No. 50-254-617 Supplier Bioworld Supplier No. 306300354
Gresshoff & Doy (DBM2) MediumContains Macronutrients, Micronutrients, and Vitamins as described Gresshoff & Doy (1974)Working Concentration: 2.71 g/L Sucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture tested