Learn More
Bioworld Gresshoff and Doy (DBM2) Medium, 25 L
Catalog No. 50254613
Encompass_Preferred
Gresshoff & Doy (DBM2) MediumContains Macronutrients and Micronutrients as described Gresshoff & Doy (1974) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested
Catalog No. 50-254-613 Supplier Bioworld Supplier No. 306300343
Gresshoff & Doy (DBM2) MediumContains Macronutrients and Micronutrients as described Gresshoff & Doy (1974) Sucrose, Cytokines, Vitamins, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of medium Plant tissue culture tested