Learn More
Bioworld Gresshoff and Doy (DBM2) Vitamin Mix, 100 mL
Supplier:  Bioworld 306300321
Encompass_Preferred
Gresshof & Doy (DBM2) Vitamin MixContains Vitamins as described Gresshoff & Doy (1974)Package contains 11.60 g vitamins to prepare 100 mL of 1000X vitamin stock Use 1 mL per liter of medium.Contents: Glycine: 4.00 mg/L myo-Inositol: 100.00 mg/L Nicotinic Acid: 1.00 mg/L Pyridoxine HCl: 1.00 mg/LThiamine HCl: 10.00 mg/L
Catalog No. 50-254-610