Learn More
Bellco Glass, Inc. Grey Open Top S/Cap,45mm With 1 5/16 Hole"
SDP
Catalog No. 501945752
Encompass_Preferred
Ideal for addition & withdraw of contents via syringe. 45mm cap has 33mm hole to accommodate 45mm septa or 45mm port assembly. 32mm cap has 20mm hole to accommodate 32mm septa or 32mm port assembly. MADE IN THE USA Polypropylene Size: 45mm Color: Grey
Catalog No. 50-194-5752 Supplier Bellco Glass, Inc. Supplier No. 563700025
Ideal for addition & withdraw of contents via syringe. 45mm cap has 33mm hole to accommodate 45mm septa or 45mm port assembly. 32mm cap has 20mm hole to accommodate 32mm septa or 32mm port assembly. MADE IN THE USA Polypropylene Size: 45mm Color: Grey