Learn More
Bachem GRF (1-29) amide (human) Acetate
SDP
Supplier:  Bachem 4030689.0001
Encompass_Preferred
GRF (1-29) amide (human) Acetate, H-Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2, 86168-78-7, C149H246N44O42S, Mr 3357.93, 1mg. Sermorelin. GRF (1-29) amide (sermorelin) is the shortest GRF fragment with full biological activity. CAS Number (sermorelin acetate): 114466-38-5.
Catalog No. 50-194-6990