Learn More
Echelon Biosciences Research Labs Growth Hormone Releasing Factor(1-44), porcine; GRF (1-44), porcine - 1 mg, CAS#: 88384-73-0
SDP
Supplier:  Echelon Biosciences Research Labs 534291MG
Encompass_Preferred
Growth Hormone Releasing Factor(1-44), porcine; GRF (1-44), porcine - 1mg , Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-Gln-Gln-Gly-Glu-Arg-Asn-Gln-Glu-Gln-Gly-Ala-Arg-Val-Arg-Leu-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8595