Learn More
Echelon Biosciences Research Labs GRF [Ac-Tyr1,D-Arg2] (1-29) amide - 0.5 mg,
SDP
Supplier:  Echelon Biosciences Research Labs 531840.5MG
Encompass_Preferred
GRF [Ac-Tyr1,D-Arg2] (1-29) amide - 0.5mg , Ac-Tyr-D-Arg-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2 Category/Application: Neuropeptides Hormones This item is non returnable
Catalog No. 50-190-8586