Learn More
Bachem GRF (human) Acetate
SDP
Supplier:  Bachem 4011472.0001
Encompass_Preferred
GRF (human) Acetate, H-Tyr-Ala-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-Gln-Gln-Gly-Glu-Ser-Asn-Gln-Glu-Arg-Gly-Ala-Arg-Ala-Arg-Leu-NH2, 83930-13-6, C215H358N72O66S, Mr 5039.72, 1mg. Growth Hormone-Releasing Factor (human), Somatorelin, Growth Hormone-Releasing Hormone (human), GHRH (human), Somatoliberin (human), Somatocrinin (human), GHRF (1-44), human. GRF is the hypothalamic peptide hormone that specifically stimulates synthesis and release of the growth hormone by somatotropic cells of the anterior pituitary gland.
Catalog No. 50-194-6992