Learn More
Medchemexpress LLC GRGDSP TFA | 98.0% | 701.61 | 25 MG
SDP
Catalog No. 5000426298
Encompass_Preferred
GRGDSP (TFA) is an integrin inhibitor. This synthetic linear RGD peptide can inhibit the adherence of tumor cells to endothelial cells of blood vessels, thereby limiting metastasis. It is also utilized to modify the surface of cardiovascular implants, such as vascular grafts, to promote endothelialization, and is widely employed in integrin research as a soluble integrin-blocking RGD-based peptide.
  • Inhibits adherence of tumor cells to endothelial cells of blood vessels
  • Limits metastasis
  • Promotes endothelialization for cardiovascular implants
  • Used in integrin research
  • Functions as a soluble integrin-blocking RGD-based peptide
Catalog No. 50-004-26298 Supplier Medchemexpress LLC Supplier No. 25MGHYP0290A
GRGDSP (TFA) is an integrin inhibitor. This synthetic linear RGD peptide can inhibit the adherence of tumor cells to endothelial cells of blood vessels, thereby limiting metastasis. It is also utilized to modify the surface of cardiovascular implants, such as vascular grafts, to promote endothelialization, and is widely employed in integrin research as a soluble integrin-blocking RGD-based peptide.
  • Inhibits adherence of tumor cells to endothelial cells of blood vessels
  • Limits metastasis
  • Promotes endothelialization for cardiovascular implants
  • Used in integrin research
  • Functions as a soluble integrin-blocking RGD-based peptide