Learn More
Medchemexpress LLC GRGDSPC TFA | 99.2% | 804.75 | 5 MG
SDP
Catalog No. 5000422232
Encompass_Preferred
GRGDSPC TFA is a 7-amino acid thiolated cell adhesion peptide. It is utilized in conjugation reactions with acrylated dextran to regulate the interactions of human mesenchymal stem cells (hMSCs) with photocrosslinkable hydrogels.
  • Thiolated cell adhesion peptide
  • Composed of 7 amino acids
  • Regulates human mesenchymal stem cell interactions
  • Suitable for research applications
Catalog No. 50-004-22232 Supplier Medchemexpress LLC Supplier No. HYP1559A5MG
GRGDSPC TFA is a 7-amino acid thiolated cell adhesion peptide. It is utilized in conjugation reactions with acrylated dextran to regulate the interactions of human mesenchymal stem cells (hMSCs) with photocrosslinkable hydrogels.
  • Thiolated cell adhesion peptide
  • Composed of 7 amino acids
  • Regulates human mesenchymal stem cell interactions
  • Suitable for research applications