Learn More
Aviva Systems Biology Corp GRHL1 Antibody - N-terminal region (ARP31917_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31917_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GRHL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GRHL1. Alternate names: LBP-32, LBP32, MGR, TFCP2L2, NH32ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54371