Learn More
Aviva Systems Biology Corp Gria1 Antibody - N-terminal region (ARP37670_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37670_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gria1. It was validated on Western Blot. . Alternate names: 2900051M01Rik, AI853806, Glr-1, Glr1, GluR-A, GluRA, Glur-1, Glur1, HIPA1, GluA1, gluR-K1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-684