Learn More
Aviva Systems Biology Corp GRIA2 Antibody - N-terminal region (AVARP13038_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13038_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GRIA2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GRIA2. Alternate names: GLUR2, GLURB, GluA2, HBGR2, GluR-K2ApplicationIHC, WB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-62620