Learn More
Aviva Systems Biology Corp Gria3 Antibody - N-terminal region (ARP35200_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35200_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Gria3. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 2900064I19Rik, GluR-C, GluR-K3, Glur-3, Glur3, Gluralpha3, KIAA4184, mKIAA4184, GluA3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-284-453