Learn More
Mitegen Grid Clipping Tool By Nanosoft, Autogrid Compatible, for Cryo-EM Sample Clipping
SDP
Catalog No. 5000495008
Encompass
The Nanosoft Grid Clipping Tool is a highly effective, compact tool designed for use with c-clips and autogrids. It is compatible with the Thermo Fisher clipping station but offers a significant advantage in its compact design. Unlike traditional versions, the Nanosoft tool is deep enough to be fully immersed in liquid nitrogen within the clipping station, preventing frost and ice contamination from environmental moisture, even during prolonged use. This allows you to pre-load multiple tools with C-clips and cool them simultaneously, enabling you to clip all your grids at once with little concern for ice. Its unique design eliminates the need to warm, re-load, and re-cool tools between clippings, streamlining your cryo-EM sample preparation workflow.
Catalog No. 50-004-95008 Supplier Mitegen Supplier No. MCEMNSCLT01
The Nanosoft Grid Clipping Tool is a highly effective, compact tool designed for use with c-clips and autogrids. It is compatible with the Thermo Fisher clipping station but offers a significant advantage in its compact design. Unlike traditional versions, the Nanosoft tool is deep enough to be fully immersed in liquid nitrogen within the clipping station, preventing frost and ice contamination from environmental moisture, even during prolonged use. This allows you to pre-load multiple tools with C-clips and cool them simultaneously, enabling you to clip all your grids at once with little concern for ice. Its unique design eliminates the need to warm, re-load, and re-cool tools between clippings, streamlining your cryo-EM sample preparation workflow.